Anti-AXL Rabbit Polyclonal Antibody
Catalog # ABGEAP16023A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:AXL Receptor Tyrosine Kinase
- Antigen symbol:AXL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:N-Terminal
- Gene ID:26362
- Antigen synonyms:Ufo|Tyro7|Ark|AI323647
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Molecular weight:98.191 kDa
- Sequence:EEPEDKAVPANTPC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,4 mg/ml
- Shipping temperature:4 °C
- Immunogen:This Mouse Axl antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 122-149 amino acids from the N-terminal region of mouse Axl.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: AXL (AXL receptor tyrosine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse