Anti-YBX3 Rabbit Polyclonal Antibody
Catalog # ABGEAP14659C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Y box binding protein 3
- Antigen symbol:YBX3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:Center
- Gene ID:8531
- Antigen synonyms:ZONAB|CSDA|CSDA1|DBPA
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Molecular weight:40.09 kDa
- Sequence:GSRYAADRRRYRRGYC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This CSDA antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 152-181 amino acids from the Central region of human CSDA.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: YBX3 (Y box binding protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse