Anti-SUPT5H Rabbit Polyclonal Antibody
ABGEAP14533C
: Abgent
- Antibody type:Primary
- Antigen name:Suppressor of Ty 5 Homolog
- Antigen symbol:SUPT5H
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:Center
- Gene ID:6829
- Antigen synonyms:Tat-CT1|SPT5|SPT5H
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Molecular weight:121 kDa
- Sequence:ESTGKEREHNFQPGDC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,3 mg/ml
- Shipping temperature:4 °C
- Immunogen:This SUPT5H antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 395-424 amino acids from the Central region of human SUPT5H.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: SUPT5H (suppressor of Ty 5 homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human