Anti-SOX9 Rabbit Polyclonal Antibody
Catalog # ABGEAP1409A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:SRY (sex determining region Y)-box 9
- Antigen symbol:SOX9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Antigen synonyms:PCB
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: SOX9 (SRY (sex determining region Y)-box 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human