Anti-TSNAX Rabbit Polyclonal Antibody
Catalog # ABGEAP13947A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Translin-associated Factor X
- Antigen symbol:TSNAX
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Antigen synonyms:MINA|Mineral dust-induced gene protein 2|MDIG|MYC-induced nuclear antigen|mina53|Protein mina53
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: TSNAX (translin-associated factor X)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human