Anti-ACVR1 Rabbit Polyclonal Antibody
ABGEAP13856C
: Abgent
- Antibody type:Primary
- Antigen name:Activin A Receptor, Type 1
- Antigen symbol:ACVR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:ACTRI|ALK2|ACVR1A|TSRI|FOP|SKR1|ACVRLK2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: ACVR1 (activin A receptor, type I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human