Anti-G3BP1 Rabbit Polyclonal Antibody
Catalog # ABGEAP13785B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:G3BP Stress Granule Assembly Factor 1
- Antigen symbol:G3BP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Antigen synonyms:G3BP|Ras GTPase-activating protein-binding protein 1|ATP-dependent DNA helicase VIII|HDH-VIII|MGC111040
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: G3BP1 (GTPase activating protein (SH3 domain) binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human