Anti-SSSCA1 Rabbit Polyclonal Antibody
ABGEAP13654A
: Abgent
- Antibody type:Primary
- Antigen name:Sjogren syndrome/scleroderma autoantigen 1
- Antigen symbol:SSSCA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:N-terminal
- Antigen synonyms:p27|ALURBP
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: SSSCA1 (Sjogren syndrome/scleroderma autoantigen 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse