Anti-DHRS7B Rabbit Polyclonal Antibody
Catalog # ABGEAP13340C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Dehydrogenase/reductase (SDR family) Member 7B
- Antigen symbol:DHRS7B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:Center
- Gene ID:25979
- Antigen synonyms:MGC8916|CGI-93|SDR32C1|DKFZp566O084|Dehydrogenase/reductase SDR family member 7B|DRS7B_HUMAN.
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Molecular weight:35.119 kDa
- Sequence:RGTIMDTTVDVDKRC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,46mg/ml
- Shipping temperature:4 °C
- Immunogen:This DHRS7B antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 133-161 amino acids from the Central region of human DHRS7B.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: DHRS7B (dehydrogenase/reductase (SDR family) member 7B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human