Anti-PHACTR3 Rabbit Polyclonal Antibody
Catalog # ABGEAP12836A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Phosphatase And Actin Regulator 3
- Antigen symbol:PHACTR3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Antigen synonyms:PLA2L
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: PHACTR3 (phosphatase and actin regulator 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human