Anti-PUS1 Rabbit Polyclonal Antibody
ABGEAP12835B
: Abgent
- Antibody type:Primary
- Antigen name:Pseudouridylate synthase 1
- Antigen symbol:PUS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: PUS1 (pseudouridylate synthase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human