Order Entry
Export
ContactUsLinkComponent
 
Anti-SPAG8 Rabbit Polyclonal Antibody
  ABGEAP12827B
 :  Abgent
Anti-SPAG8 Rabbit Polyclonal Antibody
  ABGEAP12827B
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Sperm Associated Antigen 8
  • Antigen symbol:
    SPAG8
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Antigen synonyms:
    CT142|HSD-1|SPAG3|hSMP-1|SMP1|CILD28|BS-84
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: SPAG8 (sperm associated antigen 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human