Order Entry
Export
ContactUsLinkComponent
 
Anti-CTTNBP2NL Rabbit Polyclonal Antibody
  ABGEAP12725A
 :  Abgent
undefined
Anti-CTTNBP2NL Rabbit Polyclonal Antibody
  ABGEAP12725A
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    T-SNARE Domain Containing 1
  • Antigen symbol:
    CTTNBP2NL
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: CTTNBP2NL (CTTNBP2 N-terminal like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human