Anti-PNKD Rabbit Polyclonal Antibody
ABGEAP12567A
: Abgent
- Antibody type:Primary
- Antigen name:Paroxysmal Nonkinesigenic Dyskinesia
- Antigen symbol:PNKD
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Antigen synonyms:Tudor domain-containing protein 1|CT41.1|TDRD1|Cancer/testis antigen 41.1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: PNKD (paroxysmal nonkinesigenic dyskinesia)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse