Anti-DRGX Rabbit Polyclonal Antibody
Catalog # ABGEAP12443B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Dorsal root ganglia homeobox
- Antigen symbol:DRGX
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: DRGX (dorsal root ganglia homeobox)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse