Order Entry
Export
ContactUsLinkComponent
Anti-VEGFA Rabbit Polyclonal Antibody
Anti-VEGFA Rabbit Polyclonal Antibody
Catalog # ABGEAP12184C
Supplier:  Abgent
Anti-VEGFA Rabbit Polyclonal Antibody
Catalog # ABGEAP12184C
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Vascular endothelial growth factor A
  • Antigen symbol:
    VEGFA
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    middle
  • Antigen synonyms:
    VEGF|VEGF-A,|Vascular permeability factor|Vascular Endothelial Growth Factor|Vascular endothelial growth factor A
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: VEGFA (vascular endothelial growth factor A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human