Anti-ZNF593 Rabbit Polyclonal Antibody
Catalog # ABGEAP12079C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Zinc Finger Protein 593
- Antigen symbol:ZNF593
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:middle
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: ZNF593 (zinc finger protein 593)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse