Anti-POSTN Rabbit Polyclonal Antibody
Catalog # ABGEAP11962B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Periostin
- Antigen symbol:POSTN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Antigen synonyms:PLF|OSF-2|A630052E07Rik|Osf2|PN|AI747096
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: POSTN (periostin, osteoblast specific factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human