Anti-GHSR Rabbit Polyclonal Antibody
ABGEAP11873B
: Abgent
- Antibody type:Primary
- Antigen name:Growth hormone secretagogue receptor
- Antigen symbol:GHSR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Antigen synonyms:GHSR|growth hormone secretagogue receptor|GHSR1|GHDP
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: GHSR (growth hormone secretagogue receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse