Order Entry
Export
ContactUsLinkComponent
 
Anti-ODF3 Rabbit Polyclonal Antibody
  ABGEAP11800B
 :  Abgent
Anti-ODF3 Rabbit Polyclonal Antibody
  ABGEAP11800B
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Outer Dense Fiber Of Sperm Tails 3
  • Antigen symbol:
    ODF3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: ODF3 (outer dense fiber of sperm tails 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse