Anti-GUCY2D Rabbit Polyclonal Antibody
Catalog # ABGEAP11569C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Acyl-CoA Thioesterase 11
- Antigen symbol:GUCY2D
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:BFIT|THEA|THEM1|STARD14
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: GUCY2D (guanylate cyclase 2D, membrane (retina-specific))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human