Order Entry
Export
ContactUsLinkComponent
Anti-F11 Rabbit Polyclonal Antibody
Anti-F11 Rabbit Polyclonal Antibody
Catalog # ABGEAP11568C
Supplier:  Abgent
Anti-F11 Rabbit Polyclonal Antibody
Catalog # ABGEAP11568C
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Coagulation factor 11
  • Antigen symbol:
    F11
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    middle
  • Antigen synonyms:
    F11|Coagulation factor 11|FXI
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: F11 (coagulation factor XI)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human