Anti-CASZ1 Rabbit Polyclonal Antibody
Catalog # ABGEAP10238B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Castor Zinc Finger 1
- Antigen symbol:CASZ1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Antigen synonyms:CAS11|ZNF693|CST|SRG|dJ734G22.1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: CASZ1 (castor zinc finger 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse