Anti-POTEB Rabbit Polyclonal Antibody
Catalog # ABGEAP10032B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:POTE ankyrin domain family, member B
- Antigen symbol:POTEB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:N-Terminal ( 1–100 aa)
- Cross adsorption:No
- Antigen synonyms:GDF11|Bone morphogenetic protein 11|BMP11|GDF-11|Growth/differentiation factor 11 [Precursor]
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: POTEB (POTE ankyrin domain family, member B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse