Anti-TMEM132D Rabbit Polyclonal Antibody
Catalog # ABGEAP10011B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Transmembrane Protein 132D
- Antigen symbol:TMEM132D
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-Terminal ( 1–60 aa)
- Cross adsorption:No
- Antigen synonyms:Math4C|AKA|bHLHa6|NEUROD3|ngn1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: TMEM132D (transmembrane protein 132D)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human