Anti-LAMP2 Mouse Monoclonal Antibody
ABGEAM1851B
: Abgent
- Antibody type:Primary
- Antigen name:lysosomal-associated membrane Protein 2
- Antigen symbol:LAMP2
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human,Mouse
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: LAMP2 (lysosomal-associated membrane protein 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Mouse