Anti-C9orf170 Rabbit Polyclonal Antibody
AVIVARP69899_P050
- Antibody type:Primary
- Antigen name:Chromosome 9 open reading frame 170
- Antigen symbol:C9orf170
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:401535
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: C9orf170 (chromosome 9 open reading frame 170)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human