Order Entry
Export
ContactUsLinkComponent
Anti-FAM221B Rabbit Polyclonal Antibody
Anti-FAM221B Rabbit Polyclonal Antibody
Catalog # AVIVARP69854_P050
undefined
Anti-FAM221B Rabbit Polyclonal Antibody
Catalog # AVIVARP69854_P050

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Family with sequence similarity 221, member B
  • Antigen symbol:
    FAM221B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Cross adsorption:
    none
  • Gene ID:
    392307
  • Antigen synonyms:
    C9orf128
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: FAM221B (family with sequence similarity 221, member B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human