Anti-BPIFB6 Rabbit Polyclonal Antibody
AVIVARP67999_P050
- Antibody type:Primary
- Antigen name:BPI fold containing family B, member 6
- Antigen symbol:BPIFB6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:128859
- Antigen synonyms:LPLUNC6|BPIL3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: BPIFB6 (BPI fold containing family B, member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human