Anti-YARS2 Rabbit Polyclonal Antibody
AVIVARP67913_P050
- Antibody type:Primary
- Antigen name:Tyrosyl-tRNA Synthetase 2, Mitochondrial
- Antigen symbol:YARS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:287924
- Antigen synonyms:CGI-04|RGD1311696
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: YARS2 (tyrosyl-tRNA synthetase 2, mitochondrial)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Rat