Order Entry
Export
ContactUsLinkComponent
Anti-OR52A5 Rabbit Polyclonal Antibody
Anti-OR52A5 Rabbit Polyclonal Antibody
  AVIVARP67718_P050
undefined
Anti-OR52A5 Rabbit Polyclonal Antibody
  AVIVARP67718_P050

 

  • Antibody type:
    Primary
  • Antigen name:
    olfactory receptor, family 52, subfamily A, member 5
  • Antigen symbol:
    OR52A5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Cross adsorption:
    none
  • Gene ID:
    390054
  • Antigen synonyms:
    OR11-33
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: OR52A5 (olfactory receptor, family 52, subfamily A, member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human