Anti-FTMT Rabbit Polyclonal Antibody
AVIVARP66169_P050
- Antibody type:Primary
- Antigen name:Ferritin mitochondrial
- Antigen symbol:FTMT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:94033
- Antigen synonyms:MTF
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: FTMT (ferritin mitochondrial)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human