Order Entry
Export
ContactUsLinkComponent
 
Anti-CC2D2A Rabbit Polyclonal Antibody
Catalog # AVIVARP65635_P050
undefined
Anti-CC2D2A Rabbit Polyclonal Antibody
Catalog # AVIVARP65635_P050

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    coiled-coil and C2 domain containing 2A
  • Antigen symbol:
    CC2D2A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Gene ID:
    57545
  • Antigen synonyms:
    JBTS9|MKS6
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: CC2D2A (coiled-coil and C2 domain containing 2A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human