Anti-TIMM23B Rabbit Polyclonal Antibody
AVIVARP65617_P050
- Antibody type:Primary
- Antigen name:translocase of inner mitochondrial membrane 23 homolog B (yeast)
- Antigen symbol:TIMM23B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:653252
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: TIMM23B (translocase of inner mitochondrial membrane 23 homolog B (yeast))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human