Anti-EPB41L4B Rabbit Polyclonal Antibody
Catalog # AVIVARP65563_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Erythrocyte membrane protein band 4.1 like 4B
- Antigen symbol:EPB41L4B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:54566
- Antigen synonyms:EHM2|CG1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: EPB41L4B (erythrocyte membrane protein band 4.1 like 4B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human