Anti-KRT6A Rabbit Polyclonal Antibody
Catalog # AVIVARP64418_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:keratin 6A, type II
- Antigen symbol:KRT6A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:3853
- Antigen synonyms:KRT6D|KRT6C|PC3|CK-6E|K6D|K6C|CK6A|CK6C|CK6D|CK-6C|K6A
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: KRT6A (keratin 6A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human