Anti-UBL4A Rabbit Polyclonal Antibody
Catalog # AVIVARP59393_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:ubiquitin-like 4A
- Antigen symbol:UBL4A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:8266
- Antigen synonyms:GDX|DX254E|G6PD|MDY2|DXS254E|GET5|UBL4|TMA24
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: UBL4A (ubiquitin-like 4A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: