Order Entry
Export
ContactUsLinkComponent
 
Anti-TAF1C Rabbit Polyclonal Antibody
  AVIVARP57920_P050
undefined
Anti-TAF1C Rabbit Polyclonal Antibody
  AVIVARP57920_P050

 

  • Antibody type:
    Primary
  • Antigen name:
    TATA box binding protein (TBP)-associated factor, RNA polymerase I, C, 110kDa
  • Antigen symbol:
    TAF1C
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Gene ID:
    9013
  • Antigen synonyms:
    MGC:39976|TAFI110|SL1|TAFI95
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: TAF1C (TATA box binding protein (TBP)-associated factor, RNA polymerase I, C, 110kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: