Anti-ATG4B Rabbit Polyclonal Antibody
AVIVARP57862_P050
- Antibody type:Primary
- Antigen name:autophagy related 4B, cysteine peptidase
- Antigen symbol:ATG4B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:23192
- Antigen synonyms:APG4B|AUTL1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: ATG4B (autophagy related 4B, cysteine peptidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: