Anti-MPRIP Rabbit Polyclonal Antibody
Catalog # AVIVARP57861_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Myosin Phosphatase Rho Interacting Protein
- Antigen symbol:MPRIP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:116504
- Antigen synonyms:p116Rip|Rhoip3|Mrip|M-rip
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: MPRIP (myosin phosphatase Rho interacting protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Rat