Anti-MLST8 Rabbit Polyclonal Antibody
Catalog # AVIVARP57630_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:MTOR associated protein, LST8 homolog
- Antigen symbol:MLST8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:64223
- Antigen synonyms:POP3|GbetaL|GBL|LST8|WAT1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: MLST8 (MTOR associated protein, LST8 homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human