Order Entry
Export
ContactUsLinkComponent
Anti-NECAB1 Rabbit Polyclonal Antibody
Anti-NECAB1 Rabbit Polyclonal Antibody
Catalog # AVIVARP57628_P050
undefined
Anti-NECAB1 Rabbit Polyclonal Antibody
Catalog # AVIVARP57628_P050

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    N-terminal EF-hand calcium binding protein 1
  • Antigen symbol:
    NECAB1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Cross adsorption:
    none
  • Gene ID:
    69352
  • Antigen synonyms:
    1700003H21Rik|Efcbp1|STIP-1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: NECAB1 (N-terminal EF-hand calcium binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: