Anti-SMEK2 Rabbit Polyclonal Antibody
Catalog # AVIVARP57424_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:SMEK homolog 2, suppressor of mek1 (Dictyostelium)
- Antigen symbol:SMEK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:104570
- Antigen synonyms:AW557776|Ppp4r3b|mKIAA1387|AW011752
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: SMEK2 (SMEK homolog 2, suppressor of mek1 (Dictyostelium))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: