Anti-TBC1D1 Rabbit Polyclonal Antibody
Catalog # AVIVARP55189_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:TBC1 (tre-2/USP6, BUB2, cdc16) domain family, member 1
- Antigen symbol:TBC1D1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:23216
- Antigen synonyms:TBC1|TBC
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: TBC1D1 (TBC1 (tre-2/USP6, BUB2, cdc16) domain family, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: