Anti-CDK5 Rabbit Polyclonal Antibody
AVIVARP54260_P050
- Antibody type:Primary
- Antigen name:cyclin-dependent kinase 5
- Antigen symbol:CDK5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:140908
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: CDK5 (cyclin-dependent kinase 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Rat