Order Entry
Export
ContactUsLinkComponent
Anti-WDR49 Rabbit Polyclonal Antibody
Anti-WDR49 Rabbit Polyclonal Antibody
  AVIVARP53376_P050
undefined
Anti-WDR49 Rabbit Polyclonal Antibody
  AVIVARP53376_P050

 

  • Antibody type:
    Primary
  • Antigen name:
    WD repeat domain 49
  • Antigen symbol:
    WDR49
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Gene ID:
    151790
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: WDR49 (WD repeat domain 49)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: