Anti-PTH2 Rabbit Polyclonal Antibody
AVIVARP53357_P050
- Antibody type:Primary
- Antigen name:parathyroid hormone 2
- Antigen symbol:PTH2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:113091
- Antigen synonyms:TIP39
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: PTH2 (parathyroid hormone 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: