Anti-CCDC50 Rabbit Polyclonal Antibody
AVIVARP53356_P050
- Antibody type:Primary
- Antigen name:coiled-coil domain containing 50
- Antigen symbol:CCDC50
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:152137
- Antigen synonyms:YMER|DFNA44|C3orf6
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: CCDC50 (coiled-coil domain containing 50)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: