Anti-NLGN4X Rabbit Polyclonal Antibody
Catalog # AVIVARP49557_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Neuroligin 4, X-linked
- Antigen symbol:NLGN4X
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:57502
- Antigen synonyms:ASPGX2, AUTSX2, HLNX, HNL4X, NLGN4
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: NLGN4X (neuroligin 4, X-linked)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: