Anti-JPH1 Rabbit Polyclonal Antibody
Catalog # AVIVARP49532_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Junctophilin 1
- Antigen symbol:JPH1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:56704
- Antigen synonyms:JP1|JP-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: JPH1 (junctophilin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: